Isobutylbenzene

Supplier: Matrix Scientific

Synonyms: 1-Phenylisobutane

057184-25G
101912-674EA 138.46 USD
101912-674
Isobutylbenzene
Isobutylbenzene
Formula: (C₆H₅)CH₂CH(CH₃)CH₃
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –51 °C
Density: 0.8538 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008936
CAS Number: 538-93-2
EINECS: 208-706-2
UN: 2709
ADR: 3,III
Merck Index: 13,05151

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR