Heptadecylbenzene

Supplier: Matrix Scientific

Synonyms: 1-Phenylheptadecane

057183-25G
101912-670EA 258.46 USD
101912-670
Heptadecylbenzene
Heptadecylbenzene
Formula: C₂₃H₄₀
MW: 316.57 g/mol
Boiling Pt: 397 °C (1013 hPa)
Melting Pt: 32 °C
Density: 0.8546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039873
CAS Number: 14752-75-1
EINECS: 238-812-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR