4-Phenoxybenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

057171-50G
101912-626EA 307.69 USD
101912-626
4-Phenoxybenzoic acid ≥98%
4-Phenoxybenzoic acid

Hazard code(s): Irritant

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Melting Pt: 163…165 °C
MDL Number: MFCD00002539
CAS Number: 2215-77-2
EINECS: 218-682-5

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR