5-Nitroanthranilic acid tech

Supplier: Matrix Scientific
Warning

Synonyms: 2-Amino-5-nitrobenzoic acid

057157-50G
101912-570EA 1015.38 USD
101912-570
5-Nitroanthranilic acid tech
5-Nitroanthranilic acid

Hazard code(s): Irritant

Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Melting Pt: 268…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00017039
CAS Number: 616-79-5
EINECS: 210-493-6

Order Now

Specification Test Results

Melting point 268-270°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR