1-Methyl-2-thiourea ≥98%

Supplier: Matrix Scientific

057153-50G
101912-554EA 183.08 USD
101912-554
1-Methyl-2-thiourea ≥98%
1-Methyl-2-thiourea

Hazard code(s): Irritant

Formula: C₂H₆N₂S
MW: 90.15 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD00004938
CAS Number: 598-52-7
EINECS: 209-936-6
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR