1-(4-Methylphenyl)urea

Supplier: Matrix Scientific

Synonyms: p-Tolylurea4-Methylphenylurea1-(4-Tolyl)ureaN-(4-Methylphenyl)urea1-(p-Tolyl)urea

057147-10G
101912-534EA 207.69 USD
101912-534
1-(4-Methylphenyl)urea
1-(4-Methylphenyl)urea

Hazard code(s): Irritant

Formula: (C₆H₄(CH₃))NC(O)NH₂
MW: 150.18 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 182…183 °C
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00025433
CAS Number: 622-51-5
EINECS: 210-739-2

Order Now

Specification Test Results

Melting point 182-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR