2-Methylbutyl isothiocyanate ≥98%

Supplier: Matrix Scientific

057134-5G
101912-490EA 315.38 USD
101912-490
2-Methylbutyl isothiocyanate ≥98%
2-Methylbutyl isothiocyanate

Hazard code(s): Irritant, LACHRYMATOR

Formula: C₆H₁₁NS
MW: 129.23 g/mol
Boiling Pt: 64 °C (1mm)
Density: 0.95 g/cm³
MDL Number: MFCD00060414
CAS Number: 4404-51-7

Order Now

Specification Test Results

Boiling point 64°C/1mm
Density 0.95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR