1-Methylbenzotriazole
Supplier: Matrix Scientific
Synonyms:
1-Methyl-1H-benzotriazole
057129-50G
101912-470EA
730.77
USD
101912-470
1-Methylbenzotriazole
1-Methylbenzotriazole
Hazard code(s): Irritant
Formula:
C₇H₇N₃ Melting Pt: 64…65 °C |
MDL Number:
MFCD00014572 CAS Number: 13351-73-0 EINECS: 236-401-4 |
Specification Test Results
Melting point | 64-65°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...