1-(4-Methoxyphenyl)-2-thiourea

Supplier: Matrix Scientific
Danger

Synonyms: N-(4-Methoxyphenyl)thiourea(4-Methoxyphenyl)thiourea4-Methoxyphenylthiourea1-(4-Methoxyphenyl)thiourea

057128-50G
101912-466EA 632.31 USD
101912-466
1-(4-Methoxyphenyl)-2-thiourea
1-(4-Methoxyphenyl)-2-thiourea

Hazard code(s): Irritant

Formula: (C₆H₄(OCH₃))NC(S)NH₂
MW: 182.25 g/mol
Melting Pt: 212 °C (dec.)
MDL Number: MFCD00004936
CAS Number: 2293-07-4
EINECS: 218-931-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 212°C (dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR