4-Isopropylbenzonitrile

057109-25G
101912-408EA 1035.61 USD
101912-408
4-Isopropylbenzonitrile
4-Isopropylbenzonitrile

Hazard code(s): Irritant

Formula: C₁₀H₁₁N
MW: 145.2 g/mol
Boiling Pt: 88…90 °C (5mm)
MDL Number: MFCD00052966
CAS Number: 13816-33-6
EINECS: 237-492-3

Order Now

Specification Test Results

Boiling point 88-90°C/5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR