4-Ethylbenzaldehyde ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: p-Ethylbenzaldehyde

057069-250G
101912-246EA 269.23 USD
101912-246
4-Ethylbenzaldehyde ≥99%
4-Ethylbenzaldehyde

Hazard code(s): Irritant, Refrigerate

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 220…221 °C
Density: 0.98 g/cm³
MDL Number: MFCD00006956
CAS Number: 4748-78-1
EINECS: 225-268-8

Order Now

Specification Test Results

Boiling point 220-221°C
Density 0.98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR