(2,4-Dimethylphenyl)acetonitrile ≥99%
Supplier: Matrix Scientific
057039-5G
057039-25G
101912-136EA
632.31
USD
101912-136
101912-134
(2,4-Dimethylphenyl)acetonitrile ≥99%
(2,4-Dimethylphenyl)acetonitrile
Hazard code(s): Irritant
Formula:
C₁₀H₁₁N Boiling Pt: 85 °C (1mm) Density: 0.99 g/cm³ |
MDL Number:
MFCD00060298 CAS Number: 68429-53-8 |
Specification Test Results
Boiling point | 85°C/1mm |
Density | 0.99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...