2,6-Diiodo-4-nitroaniline
Supplier: Matrix Scientific
|
Warning
|
057022-25G
057022-5G
101912-076EA
130.77
USD
101912-076
101912-078
2,6-Diiodo-4-nitroaniline
2,6-Diiodo-4-nitroaniline
Hazard code(s): Irritant
Formula:
C₆H₄I₂N₂O₂ MW: 389.92 g/mol Melting Pt: 243…245 °C |
MDL Number:
MFCD00007683 CAS Number: 5398-27-6 EINECS: 226-429-5 |
Specification Test Results
Melting point | 243-245°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...