1-(3-Chlorophenyl)-2-thiourea ≥98%
Supplier: Matrix Scientific
|
Danger
|
Synonyms:
(3-Chlorophenyl)thiourea
, N-(3-Chlorophenyl)thiourea
, 3-Chlorophenylthiourea
056983-50G
056983-10G
101911-956EA
223.08
USD
101911-956
101911-958
1-(3-Chlorophenyl)-2-thiourea ≥98%
1-(3-Chlorophenyl)-2-thiourea
Hazard code(s): Irritant
Formula:
(C₆H₄Cl)NC(S)NH₂ MW: 186.66 g/mol Boiling Pt: 304 °C (1013 hPa) Melting Pt: 138 °C |
MDL Number:
MFCD00022165 CAS Number: 4947-89-1 UN: 2811 ADR: 6.1,II |
Specification Test Results
Melting point | 138°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...