3-Chlorobenzamide

Supplier: Matrix Scientific

056945-10G 056945-50G
101911-820EA 307.69 USD
101911-820 101911-818
3-Chlorobenzamide
3-Chlorobenzamide

Hazard code(s): Irritant

Formula: C₇H₆ClNO
MW: 155.58 g/mol
Melting Pt: 134…135 °C
MDL Number: MFCD00014799
CAS Number: 618-48-4
EINECS: 210-554-7

Order Now

Specification Test Results

Melting point 134-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR