2-Bromo-m-xylene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2-Bromo-1,3-dimethylbenzene2,6-Dimethylbromobenzene

056934-25G
101911-788EA 29.23 USD
101911-788
2-Bromo-m-xylene ≥98%
2-Bromo-m-xylene

Hazard code(s): Irritant

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 206 °C
Melting Pt: –10 °C
Density: 1.39 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000075
CAS Number: 576-22-7
EINECS: 209-397-7

Order Now

Specification Test Results

Boiling point 206°C
Density 1.39

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR