4-Bromosalicylic acid

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-hydroxybenzoic acid

056913-25G
101911-704EA 106.15 USD
101911-704
4-Bromosalicylic acid
4-Bromosalicylic acid

Hazard code(s): Irritant

Formula: C₇H₅BrO₃
MW: 217.02 g/mol
Melting Pt: 214 °C
MDL Number: MFCD06797970
CAS Number: 1666-28-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR