Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Supplier: Matrix Scientific
Synonyms:
Benzo-18-crown 6-Ether
056891-5G
056891-1G
101911-612EA
201.54
USD
101911-612
101911-614
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Benzo-18-crown-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Hazard code(s): Irritant
Formula:
C₁₆H₂₄O₆ MW: 312.36 g/mol Melting Pt: 40…42 °C |
MDL Number:
MFCD00062741 CAS Number: 14098-24-9 |
Specification Test Results
Melting point | 40-42°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...