4-Bromo-3,5-dimethylanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-methoxy-1,3-dimethylbenzene

056849-5G
101911-528EA 29.23 USD
101911-528
4-Bromo-3,5-dimethylanisole ≥98%
4-Bromo-3,5-dimethylanisole

Hazard code(s): Irritant

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
MDL Number: MFCD00461692
CAS Number: 6267-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR