2,6-Dichlorothiobenzamide ≥98%

Supplier: Matrix Scientific

Synonyms: Chlorthiamid

056839-1G
101911-506EA 106.15 USD
101911-506
2,6-Dichlorothiobenzamide ≥98%
2,6-Dichlorothiobenzamide

Hazard code(s): Irritant

Formula: C₇H₅Cl₂NS
MW: 206.09 g/mol
MDL Number: MFCD00053523
CAS Number: 1918-13-4
EINECS: 217-637-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR