1-(2,4,5-Trichlorophenyl)-2-thiourea

Supplier: Matrix Scientific

056831-1G
101911-490EA 56.92 USD
101911-490
1-(2,4,5-Trichlorophenyl)-2-thiourea
1-(2,4,5-Trichlorophenyl)-2-thiourea

Hazard code(s): Irritant

Formula: (C₆H₂Cl₃)NC(S)NH₂
MW: 255.55 g/mol
Melting Pt: 178…180 °C
MDL Number: MFCD00060431
CAS Number: 90617-76-8

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR