N-(3-Pyridyl)thiourea ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(3-Pyridyl)-2-thiourea

056824-5G
101911-476EA 226.15 USD
101911-476
N-(3-Pyridyl)thiourea ≥98%
N-(3-Pyridyl)thiourea

Hazard code(s): Irritant

Formula: C₆H₇N₃S
Melting Pt: 159…160 °C
MDL Number: MFCD00014635
CAS Number: 30162-37-9
EINECS: 250-075-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 159-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR