N-(2-Pyridyl)thiourea ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(Pyridin-2-yl)thiourea
, (2-Pyridyl)thiourea
, 1-(2-Pyridyl)-2-thiourea
056823-5G
101911-474EA
80
USD
101911-474
N-(2-Pyridyl)thiourea ≥97%
N-(2-Pyridyl)thiourea
Hazard code(s): Irritant
Formula:
C₆H₇N₃S MW: 153.21 g/mol Melting Pt: 148…150 °C |
MDL Number:
MFCD00041227 CAS Number: 14294-11-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 148-150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...