Octadecylbenzene

Supplier: Matrix Scientific

Synonyms: 1-Phenyloctadecane

056809-5G
101911-444EA 229.23 USD
101911-444
Octadecylbenzene
Octadecylbenzene

Hazard code(s): Irritant

Formula: C₂₄H₄₂
MW: 330.6 g/mol
Boiling Pt: 408 °C (1013 hPa)
Melting Pt: 34…36 °C
Density: 0.8546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00048500
CAS Number: 4445-07-2
EINECS: 224-684-7

Order Now

Specification Test Results

Melting point 34-36°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR