Phenyl anthranilate ≥98%

Supplier: Matrix Scientific

056805-10G
101911-436EA 215.38 USD
101911-436
Phenyl anthranilate ≥98%
Phenyl anthranilate

Hazard code(s): Irritant

Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00060603
CAS Number: 10268-69-6
EINECS: 233-608-1

Order Now

Specification Test Results

Melting point 70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR