Phenylacetamide ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Phenylacetamide

056800-25G
101911-426EA 81.54 USD
101911-426
Phenylacetamide ≥99%
Phenylacetamide

Hazard code(s): Irritant-Harmful

Formula: C₈H₉NO
MW: 135.17 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00059193
CAS Number: 103-81-1
EINECS: 203-147-0

Order Now

Specification Test Results

Melting point 156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR