1-(2-Methylphenyl)-2-thiourea

Supplier: Matrix Scientific
Danger

Synonyms: N-(2-Methylphenyl)thiourea1-(2-Tolyl)thioureaN-(o-Tolyl)thiourea2-Methylphenylthioureao-Tolylthiourea

056768-5G
101911-362EA 44.62 USD
101911-362
1-(2-Methylphenyl)-2-thiourea
1-(2-Methylphenyl)-2-thiourea

Hazard code(s): Irritant

Formula: (C₆H₄(CH₃))NC(S)NH₂
MW: 166.25 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 158…161 °C
MDL Number: MFCD00041162
CAS Number: 614-78-8
EINECS: 210-395-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 158-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR