Methyl-3,4-diaminobenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3,4-diaminobenzoate3,4-Diaminobenzoic acid methyl ester3,4-Diamino benzoate

056761-5G 056761-25G
101911-348EA 90.77 USD
101911-348 101911-346
Methyl-3,4-diaminobenzoate ≥97%
Methyl 3,4-diaminobenzoate

Hazard code(s): Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 107…109 °C
MDL Number: MFCD00017098
CAS Number: 36692-49-6

Order Now

Specification Test Results

Melting point 107-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR