α-Bromo-3-iodotoulene ≥98%

Supplier: Matrix Scientific
Danger

Synonyms: α-Bromo-m-iodotoulene3-Iodobenzyl bromide1-(Bromomethyl)-3-iodobenzene

056742-5G
101911-308EA 110.77 USD
101911-308
α-Bromo-3-iodotoulene ≥98%
α-Bromo-3-iodotoulene

Hazard code(s): Corrosive, Lachrymator, Refrigerate

Formula: C₇H₆BrI
MW: 296.93 g/mol
Melting Pt: 49…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00019016
CAS Number: 49617-83-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 49-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR