4-Ethoxybenzonitrile ≥99%

Supplier: Matrix Scientific

Synonyms: p-Ethoxybenzonitrile

056723-10G
101911-272EA 158.46 USD
101911-272
4-Ethoxybenzonitrile ≥99%
4-Ethoxybenzonitrile

Hazard code(s): Irritant

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 258 °C
Melting Pt: 62…64 °C
MDL Number: MFCD00001819
CAS Number: 25117-74-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 258°C
Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR