(3,5-Dinitro-4-hydroxyphenyl)acetic acid

Supplier: Matrix Scientific

056721-5G
101911-268EA 158.46 USD
101911-268
(3,5-Dinitro-4-hydroxyphenyl)acetic acid
(3,5-Dinitro-4-hydroxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: C₈H₆N₂O₇
MW: 242.14 g/mol
Melting Pt: 166…168 °C
MDL Number: MFCD00016995
CAS Number: 10463-37-3

Order Now

Specification Test Results

Melting point 166-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR