1-(2,4-Dimethoxyphenyl)-2-thiourea

Supplier: Matrix Scientific

Synonyms: 2,4-DimethoxyphenylthioureaN-(2,4-Dimethoxyphenyl)thiourea

056716-5G
101911-258EA 60 USD
101911-258
1-(2,4-Dimethoxyphenyl)-2-thiourea
1-(2,4-Dimethoxyphenyl)-2-thiourea

Hazard code(s): Irritant

Formula: (C₆H₃(OCH₃)₂)NC(S)NH₂
MW: 212.27 g/mol
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 199…201 °C
MDL Number: MFCD00041158
CAS Number: 35696-77-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR