1-tert-Butyl-2-thiourea

Supplier: Matrix Scientific

Synonyms: N-(tert-Butyl)thiourea

056669-5G
101911-168EA 127.65 USD
101911-168
1-tert-Butyl-2-thiourea
1-tert-Butyl-2-thiourea

Hazard code(s): Irritant

Formula: C₅H₁₂N₂S
Melting Pt: 171…172 °C
MDL Number: MFCD00041192
CAS Number: 7204-48-0

Order Now

Specification Test Results

Melting point 171-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR