Benzylthiourea ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Benzyl-2-thioureaN-Benzylthiourea

056653-5G
101911-138EA 43.08 USD
101911-138
Benzylthiourea ≥98%
Benzylthiourea

Hazard code(s): Toxic, Irritant

Formula: C₈H₁₀N₂S
Melting Pt: 162…166 °C
MDL Number: MFCD00041370
CAS Number: 621-83-0
EINECS: 210-709-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR