N-(10-Bromodecyl)phthalimide ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(10-Bromodecyl)isoindoline-1,3-dione

056632-5G
101911-094EA 241.54 USD
101911-094
N-(10-Bromodecyl)phthalimide ≥98%
N-(10-Bromodecyl)phthalimide

Hazard code(s): Irritant

Formula: C₁₈H₂₄BrNO₂
MW: 366.3 g/mol
Melting Pt: 58…60 °C
MDL Number: MFCD00060521
CAS Number: 24566-80-1

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR