4-Methoxybenzyl isothiocyanate ≥98%

Supplier: Matrix Scientific

Synonyms: 4-MBITC4-Methoxybenzylisothiocyanate

056577-5G
101910-936EA 423.08 USD
101910-936
4-Methoxybenzyl isothiocyanate ≥98%
4-Methoxybenzyl isothiocyanate

Hazard code(s): Irritant-Harmful

Formula: C₉H₉NOS
MW: 179.24 g/mol
Boiling Pt: 133 °C (0.7mm)
Density: 1.15 g/cm³
MDL Number: MFCD00041117
CAS Number: 3694-57-3

Order Now

Specification Test Results

Boiling point 133°C/0.7mm
Density 1.15

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR