3-Methoxybenzyl isothiocyanate ≥99%
Supplier: Matrix Scientific
056576-1G
056576-5G
101910-934EA
289.23
USD
101910-934
101910-932
3-Methoxybenzyl isothiocyanate ≥99%
3-Methoxybenzyl isothiocyanate
Hazard code(s): Irritant-Harmful
Formula:
C₉H₉NOS MW: 179.24 g/mol Boiling Pt: 120 °C (1mm) Density: 1.16 g/cm³ |
MDL Number:
MFCD00060408 CAS Number: 75272-77-4 |
Specification Test Results
Boiling point | 120°C/1mm |
Density | 1.16 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...