Ethyl isothiocyanatoacetate ≥99%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-isothiocyanatoacetate

056560-5G
101910-878EA 62.1 USD
101910-878
Ethyl isothiocyanatoacetate ≥99%
Ethyl isothiocyanatoacetate

Hazard code(s): Toxic, Refrigerate

Formula: C₅H₇NO₂S
Boiling Pt: 104…106 °C (7mm)
Density: 1.17 g/cm³
MDL Number: MFCD00060677
CAS Number: 24066-82-8
EINECS: 417-720-1

Order Now

Specification Test Results

Boiling point 61°C/2mm
Density 1.17

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR