1-Decyl isothiocyanate ≥99%

Supplier: Matrix Scientific

Synonyms: Decyl isothiocyanate

056557-5G
101910-872EA 207.69 USD
101910-872
1-Decyl isothiocyanate ≥99%
1-Decyl isothiocyanate

Hazard code(s): Irritant-HARMFUL, CORROSIVE

Formula: C₁₁H₂₁NS
MW: 199.36 g/mol
Boiling Pt: 171 °C (35mm)
Density: 0.89 g/cm³
MDL Number: MFCD00041120
CAS Number: 24540-94-1

Order Now

Specification Test Results

Boiling point 171°C/35mm
Density 0.89

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR