Cyclododecyl isothiocyanate

Supplier: Matrix Scientific

056554-5G
101910-864EA 126.15 USD
101910-864
Cyclododecyl isothiocyanate
Cyclododecyl isothiocyanate

Hazard code(s): Irritant-HARMFUL

Formula: C₁₃H₂₃NS
MW: 225.4 g/mol
Boiling Pt: 163 °C (5mm)
Density: 0.99 g/cm³
MDL Number: MFCD00041029
CAS Number: 59037-64-8

Order Now

Specification Test Results

Boiling point 163°C/5mm
Density 0.99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR