2-Chloro-4-methylphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056552-5G 056552-1G
101910-858EA 36.92 USD
101910-858 101910-860
2-Chloro-4-methylphenyl isothiocyanate ≥98%
2-Chloro-4-methylphenyl isothiocyanate

Hazard code(s): Irritant-HARMFUL, LACHRYMATOR

Formula: C₈H₆ClNS
Boiling Pt: 138 °C (20mm)
Density: 1.22 g/cm³
MDL Number: MFCD00046791
CAS Number: 57878-93-0

Order Now

Specification Test Results

Boiling point 138°C/20mm
Density 1.22

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR