Methyl-2-chloro-5-nitronicotinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-5-nitronicotinate

055487-1G
101909-494EA 73.6 USD
101909-494
Methyl-2-chloro-5-nitronicotinate ≥97%
Methyl 2-chloro-5-nitronicotinate

Hazard code(s): Irritant

Formula: C₇H₅ClN₂O₄
MW: 216.58 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD07368888
CAS Number: 190271-88-6

Order Now

Specification Test Results

Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR