Tetrabromo-p-benzoquinone ≥95%
Synonyms:
Tetrabromo-1,4-benzoquinone
, Tetrabromoquinone
, 4-Bromanil
, 2,3,5,6-Tetrabromo-1,4-benzoquinone
, 2,3,5,6-Tetrabromocyclohexa-2,5-diene-1,4-dione
, p-Bromanil
055402-5G
101909-186EA
235.75
USD
101909-186
Tetrabromo-p-benzoquinone ≥95%
Tetrabromo-p-benzoquinone
Hazard code(s): Irritant
Formula:
C₆Br₄O₂ MW: 423.68 g/mol |
MDL Number:
MFCD00013785 CAS Number: 488-48-2 EINECS: 207-679-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...