2,4-Dibromopyrimidine ≥98%

Supplier: Matrix Scientific
Warning

054597-1G
101907-884EA 46.15 USD
101907-884
2,4-Dibromopyrimidine ≥98%
2,4-Dibromopyrimidine

Hazard code(s): Irritant, Refrigerate, Stored under Argon

Formula: C₄H₂Br₂N₂
MW: 237.88 g/mol
Boiling Pt: 316.6 °C
Density: 2.197 g/cm³
MDL Number: MFCD06798232
CAS Number: 3921-01-5

Order Now

Specification Test Results

Boiling point 316.6°C
Density 2.197
ND20 1.615

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR