4-Amino-3-bromopyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4-pyridylamine3-Bromopyridin-4-amine

054383-5G 054383-1G
101907-224EA 21.54 USD
101907-224 101907-222
4-Amino-3-bromopyridine ≥98%
4-Amino-3-bromopyridine

Hazard code(s): Irritant, Air Sensitive

Formula: C₅H₅BrN₂
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 65…69 °C
MDL Number: MFCD02068297
CAS Number: 13534-98-0

Order Now

Specification Test Results

Melting point 65-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR