5-Bromooxindole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1,3-dihydro-2H-indol-2-one5-Bromoindolin-2-one5-Bromo-2-oxindole

054217-5G
101906-626EA 36.92 USD
101906-626
5-Bromooxindole ≥95%
5-Bromooxindole

Hazard code(s): Irritant

Formula: C₈H₆BrNO
MW: 212.05 g/mol
Melting Pt: 220…224 °C
MDL Number: MFCD00456998
CAS Number: 20870-78-4

Order Now

Specification Test Results

Melting point 220-224°CC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR