4-Bromopicolinic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Bromopyridine-2-carboxylic acid
048494-1G
048494-5G
101884-078EA
87.69
USD
101884-078
101884-076
4-Bromopicolinic acid ≥98%
4-Bromopicolinic acid
Hazard code(s): Irritant
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Melting Pt: 172…173 °C |
MDL Number:
MFCD03426665 CAS Number: 30766-03-1 |
Specification Test Results
Melting point | 172-173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...