4-Bromo-3-chloropyridine ≥98%

Supplier: Matrix Scientific

048491-5G
101884-070EA 69.23 USD
101884-070
4-Bromo-3-chloropyridine ≥98%
4-Bromo-3-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₃BrClN
MDL Number: MFCD04112577
CAS Number: 73583-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR