5-Bromo-2-chloro-4-iodopyridine ≥95%

Supplier: Matrix Scientific

048073-1G
101882-692EA 276.92 USD
101882-692
5-Bromo-2-chloro-4-iodopyridine ≥95%
5-Bromo-2-chloro-4-iodopyridine

Hazard code(s): Irritant

Formula: C₅H₂BrClIN
MW: 318.34 g/mol
Melting Pt: 149…150 °C
MDL Number: MFCD09972189
CAS Number: 401892-47-5

Order Now

Specification Test Results

Melting point 149-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR