4'-Hydroxy-3'-nitroacetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Hydroxy-3-nitrophenyl)-1-ethanone
047515-1G
101880-408EA
232.31
USD
101880-408
4'-Hydroxy-3'-nitroacetophenone ≥95%
4'-Hydroxy-3'-nitroacetophenone
Hazard code(s): Irritant
Formula:
C₈H₇NO₄ |
MDL Number:
MFCD00017002 CAS Number: 6322-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...